Order Entry
Italy
ContactUsLinkComponent
Potassium-6-hydroxy-2-naphthalenesulphonate ≥98.0% (by HPLC, titration analysis)
Potassium-6-hydroxy-2-naphthalenesulphonate ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAN0703-25G
Supplier:  TCI
CAS Number:  
Potassium-6-hydroxy-2-naphthalenesulphonate ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAN0703-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Ion exchange titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :262.32 g/mol
EINECS:212-631-0
CAS Number:833-66-9
Formula:C₁₀H₈O₄S
MDL Number:MFCD06797149