Nω-Nitro-L-arginine methyl ester hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:51298-62-5
:
Formula:C₇H₁₅N₅O₄
MDL Number:MFCD00039052
:269.69 g/mol
EINECS:257-116-1
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Argentmetric Titration):min. 98.0 %
- Specific rotation [a]20/D:+14.0 to +16.0 deg (C=3, MeOH)
- Solubility in Water:almost transparency
Specification Test Results
Frequently Bought Together