Order Entry
Italy
ContactUsLinkComponent
3-Nonen-2-one ≥96.0% (by GC)
3-Nonen-2-one ≥96.0% (by GC)
  TCIAN0592-25ML
 :  TCI
 :  
3-Nonen-2-one ≥96.0% (by GC)
  TCIAN0592-25ML
 :  TCI
 :  

 

  • Pk:
    25 mL
  • Seq:
    2315

 

 

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 96.0 %

 

 

 :
 :107°C
 :81°C
 :140.23 g/mol
 :238-248-9
 :14309-57-0
 :C₉H₁₆O
 :MFCD00010241