Order Entry
Italy
ContactUsLinkComponent
cis-4-Nonene ≥95.0% (by GC)
Flammable
Health
cis-4-Nonene ≥95.0% (by GC)
Catalog # TCIAN0384-5ML
Supplier:  TCI
CAS Number:  
Flammable
Health
cis-4-Nonene ≥95.0% (by GC)
Catalog # TCIAN0384-5ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 mL
  • Seq:
    2210

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 95.0 %

Specification Test Results

About this item

 :
 :126.24 g/mol
Boiling Pt:147°C
Flash Pt:30°C
UN:3295
ADR:3,III
 :GHS02
 :GHS08
CAS Number:10405-84-2
Formula:C₉H₁₈
MDL Number:MFCD00067487