Disodium-1,6-naphthalenedisulphonate ≥98.0% (by HPLC, titration analysis)
TCIAN0011-1G
: TCI
:
- Pk:1 g
- Seq:3047
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Ion exchange titration):min. 98.0 %
:
:332.25 g/mol
:216-734-1
:1655-43-2
:C₁₀H₈O₆S₂
:MFCD00021522