3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT) ≥97.0% (by HPLC) for biochemical research
Supplier: TCI
Certificates
About this item
CAS Number:298-93-1
:
Formula:C₁₈H₁₆N₅BrS
MDL Number:MFCD00011964
:414.33 g/mol
EINECS:206-069-5
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Functionality test:to pass test (by Abs method)
- Solubility in hot Water:almost transparency
Specification Test Results
Frequently Bought Together