4-Methyl-4-(methylthio)-2-pentanone ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:23550-40-5
:
Formula:C₇H₁₄OS
MDL Number:MFCD00049253
:146.25 g/mol
Boiling Pt:78°C
EINECS:245-727-6
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together