
trans-4-(N-Maleimidomethyl)cyclohexane-1-carboxylic acid ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:69907-67-1
:
Formula:C₁₂H₁₅NO₄
MDL Number:MFCD11519178
:237.26 g/mol
:GHS07
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 97.0 %
- Melting point:164.0 to 168.0 deg-C
Specification Test Results
Frequently Bought Together