
1-Methylpyridinium bis(trifluoromethanesulphonyl)imide ≥98.0% (by HPLC, titration analysis)
TCIAM3199-5G
: TCI
:
- Pk:5 g
- Seq:3173
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Ion exchange titration):min. 98.0 %
- Melting point:43.0 to 47.0 deg-C
:
:374.27 g/mol
:GHS07
:742086-10-8
:C₈H₈F₆N₂O₄S₂