
1-Methylpyridinium Hexafluorophosphate ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:117029-34-2
:
Formula:C₆H₈F₆NP
:239.10 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:194.0 to 198.0 deg-C
Specification Test Results
Frequently Bought Together