N-Methyl-7H-purin-6-amine ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAM3166-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
About this item
:
Melting Pt:308°C
:149.16 g/mol
EINECS:207-137-7
CAS Number:443-72-1
Formula:C₆H₇N₅
MDL Number:MFCD00075820