Order Entry
Italy
ContactUsLinkComponent
 
trans,trans-4-Methoxy-4'-propyl-1,1'-bicyclohexyl ≥98.0% (by GC)
Catalog # TCIAM3140-5G
Supplier:  TCI
CAS Number:  
trans,trans-4-Methoxy-4'-propyl-1,1'-bicyclohexyl ≥98.0% (by GC)
Catalog # TCIAM3140-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g/mol
  • Seq:
    9225

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :238.42 g/mol
Boiling Pt:145°C
Melting Pt:9°C
CAS Number:97398-80-6
Formula:C₁₆H₃₀O
MDL Number:MFCD12761281