Order Entry
Italy
ContactUsLinkComponent
10-[2-(2-Methoxyethoxy)ethyl]-10H-phenothiazine ≥98.0% (by GC)
10-[2-(2-Methoxyethoxy)ethyl]-10H-phenothiazine ≥98.0% (by GC)
Catalog # TCIAM3068-10G
Supplier:  TCI
CAS Number:  
undefined
10-[2-(2-Methoxyethoxy)ethyl]-10H-phenothiazine ≥98.0% (by GC)
Catalog # TCIAM3068-10G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Yellow to Orange clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :301.40 g/mol
CAS Number:2098786-35-5
Formula:C₁₇H₁₉NO₂S
MDL Number:MFCD31618147