Order Entry
Italy
ContactUsLinkComponent
4-Methoxy-3-(methoxymethoxy)benzaldehyde ≥96.0% (by GC)
4-Methoxy-3-(methoxymethoxy)benzaldehyde ≥96.0% (by GC)
Catalog # TCIAM3066-1G
Supplier:  TCI
CAS Number:  
4-Methoxy-3-(methoxymethoxy)benzaldehyde ≥96.0% (by GC)
Catalog # TCIAM3066-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (GC):
    min. 96.0 %

Specification Test Results

About this item

 :
 :196.20 g/mol
Boiling Pt:130°C
Melting Pt:32°C
Storage Temperature:Refrigerator
CAS Number:5779-98-6
Formula:C₁₀H₁₂O₄