
4-Methoxy-2,3,6-trimethylbenzaldehyde ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:54344-92-2
:
Formula:C₁₁H₁₄O₂
MDL Number:MFCD00456729
:178.23 g/mol
EINECS:259-117-2
:GHS07
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:63.0 to 68.0 deg-C
Specification Test Results
Frequently Bought Together