Mono-6-(Fmoc-Gln-OtBu)-beta-CD ≥85.0% (by HPLC)
Catalog # TCIAM2978-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC) (CAD):min. 85.0 area%
Specification Test Results
About this item
:
:1541.47 g/mol
Storage Temperature:Freezer
Formula:C₆₆H₉₆N₂O₃₉