
4-Acetyl-N-methylaniline ≥98.0% (by GC, titration analysis)
Catalog # TCIAM2958-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:104.0 to 108.0 deg-C
Specification Test Results
About this item
:
:149.193 g/mol
:GHS07
CAS Number:17687-47-7
Formula:C₉H₁₁NO
MDL Number:MFCD09261160