Order Entry
Italy
ContactUsLinkComponent
 
 
1-Methyl-3,5-dinitro-1H-pyridin-2-one ≥98.0% (by GC)
  TCIAM2820-1G
 :  TCI
 :  
undefined
 
1-Methyl-3,5-dinitro-1H-pyridin-2-one ≥98.0% (by GC)
  TCIAM2820-1G
 :  TCI
 :  

 

  • Pk:
    1 g
  • Seq:
    3047

 

 

  • Appearance:
    Light orange to Yellow to Green powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    174.0 to 178.0 deg-C

 

 

 :
 :199.121 g/mol
 :GHS07
 :14150-94-8
 :C₆H₅N₃O₅
 :MFCD00456280