5-Methyl-4,5,6,7-tetrahydrothiazolo[5,4-c]pyridine-2-carboxylic acid hydrochloride ≥97.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:720720-96-7
:
Formula:C₈H₁₀N₂O₂S·HCl
MDL Number:MFCD25542379
:234.70 g/mol
Specification Test Results
- Appearance:Light orange to Yellow to Green powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (Neutralization titration):min. 94.0 %
Specification Test Results
Frequently Bought Together