
Morpholin-3-one ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:109-11-5
:
Formula:C₄H₇NO₂
MDL Number:MFCD00631009
:101.1039 g/mol
Boiling Pt:142°C
EINECS:203-647-9
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:103.0 to 107.0 deg-C
Specification Test Results
Frequently Bought Together