Mizolastine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:108612-45-9
:
Formula:C₂₄H₂₅FN₆O
MDL Number:MFCD23140913
:432.50 g/mol
Merck Index:14,06221
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:222.0 to 226.0 deg-C
Specification Test Results
Frequently Bought Together