
Methyl-DL-pyroglutamate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:54571-66-3
:
Formula:C₆H₉NO₃
MDL Number:MFCD03844651
:143.14 g/mol
Boiling Pt:135°C
EINECS:259-233-3
:GHS07
Specification Test Results
- Appearance:Colorless to Brown clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together