
4-(2-Morpholin-4-ylethoxy)phenylamine ≥98.0% (by GC, titration analysis)
: TCI
:52481-41-1
:
:C₁₂H₁₈N₂O₂
:MFCD04970981
:222.29 g/mol
:150°C
:GHS07
- Appearance:Light yellow to Yellow powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:69.0 to 73.0 deg-C
Frequently Bought Together