
5-Amino-2-methylbenzotrifluoride ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:65934-74-9
:
Formula:C₈H₈F₃N
MDL Number:MFCD01631582
:175.16 g/mol
Flash Pt:102°C
:GHS07
Specification Test Results
- Appearance:White to Yellow to Green clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Related Information
Frequently Bought Together