Order Entry
Italy
ContactUsLinkComponent
4,4'-Methylenebis(N,N-diglycidylaniline) ≥98.0% (by titrimetric analysis)
Irritant
Environment
4,4'-Methylenebis(N,N-diglycidylaniline) ≥98.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
undefined
Irritant
Environment
4,4'-Methylenebis(N,N-diglycidylaniline) ≥98.0% (by titrimetric analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Light yellow to Amber to Dark green clear liquid
  • Purity (Nonaqueous Titration):
    min. 98.0 %

Specification Test Results

Related Information

Frequently Bought Together