Order Entry
Italy
ContactUsLinkComponent
 
 
3-(Methoxydimethylsilyl)propyl acrylate ≥95.0% (by GC) stabilised
  TCIAM2525-5G
 :  TCI
 :  
undefined
 
3-(Methoxydimethylsilyl)propyl acrylate ≥95.0% (by GC) stabilised
  TCIAM2525-5G
 :  TCI
 :  

 

  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Purity (GC):
    min. 95.0 %

 

 

 :
 :202.33 g/mol
 :Stabilised with 4-methoxyphenol (MEHQ)
 :GHS07
 :Freezer
 :111918-90-2
 :C₉H₁₈O₃Si
 :MFCD00054801