
2-(Methylthio)benzo[d]oxazole ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:13673-62-6
:
Formula:C₈H₇NOS
MDL Number:MFCD00160027
:165.2100 g/mol
Boiling Pt:122°C
:GHS07
Specification Test Results
- Appearance:Light yellow to Yellow to Orange clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together