Order Entry
Italy
ContactUsLinkComponent
 
 
5-(Methylthio)thiophene-2-carboxaldehyde ≥98.0% (by GC)
  TCIAM2448-1G
 :  TCI
 :  
undefined
 
5-(Methylthio)thiophene-2-carboxaldehyde ≥98.0% (by GC)
  TCIAM2448-1G
 :  TCI
 :  

 

  • Pk:
    1 g
  • Seq:
    3047

 

 

  • Appearance:
    White to Orange to Green powder to lump
  • Purity (GC):
    min. 98.0 %

 

 

 :
 :158.23 g/mol
 :95°C
 :26°C
 :GHS07
 :Refrigerator
 :24445-35-0
 :C₆H₆OS₂
 :MFCD00173744