Order Entry
Italy
ContactUsLinkComponent
3-Methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene ≥98.0% (by GC, titration analysis)
3-Methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene ≥98.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
undefined
3-Methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene ≥98.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    58.0 to 62.0 deg-C

Specification Test Results

Related Information

Frequently Bought Together