

5-Methyl-3-hexen-2-one (contains 5-Methyl-4-hexen-2-one)
: TCI
:5166-53-0
:
:C₇H₁₂O
:MFCD00012166
:112.17 g/mol
:157°C
:48°C
:225-950-5
:1992
:3,III
:GHS02
:GHS06
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 50.0 %
Frequently Bought Together