Order Entry
Italy
ContactUsLinkComponent
 
2-Methylthio-4-pyrimidinol ≥98.0% (by HPLC, titration analysis)
 :  TCI
 
undefined
2-Methylthio-4-pyrimidinol ≥98.0% (by HPLC, titration analysis)
 :  TCI

 

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    200.0 to 204.0 deg-C

 

Frequently Bought Together