Order Entry
Italy
ContactUsLinkComponent
Mefloquine hydrochloride ≥98.0% (by HPLC)
Irritant
Mefloquine hydrochloride ≥98.0% (by HPLC)
Catalog # TCIAM2313-1G
Supplier:  TCI
CAS Number:  
undefined
Irritant
Mefloquine hydrochloride ≥98.0% (by HPLC)
Catalog # TCIAM2313-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 95.0 %

Specification Test Results

About this item

 :
 :414.78 g/mol
Merck Index:14,05801
EINECS:257-412-0
 :GHS07
CAS Number:51773-92-3
Formula:C₁₇H₁₆F₆N₂O
MDL Number:MFCD00797519