
3-(Methylamino)propionitrile ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:693-05-0
:
Formula:C₄H₈N₂
MDL Number:MFCD00001954
:84.12 g/mol
Boiling Pt:81°C
EINECS:211-740-0
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together