Order Entry
Italy
ContactUsLinkComponent
Methyl cis-11-octadecenoate ≥98.0% (by GC)
Methyl cis-11-octadecenoate ≥98.0% (by GC)
Catalog # TCIAM2308-100MG
Supplier:  TCI
CAS Number:  
undefined
Methyl cis-11-octadecenoate ≥98.0% (by GC)
Catalog # TCIAM2308-100MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :296.50 g/mol
Boiling Pt:136°C
Density:0.88
Flash Pt:62°C
Storage Temperature:Freezer
EINECS:217-714-5
CAS Number:1937-63-9
Formula:C₁₉H₃₆O₂
MDL Number:MFCD00010457