
3-(Methylaminomethyl)pyridine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:20173-04-0
:
Formula:C₇H₁₀N₂
MDL Number:MFCD00023610
Flash Pt:93°C
:122.17 g/mol
EINECS:243-561-9
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together