
4-Methoxy-2,6-dimethylphenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI
Certificates
About this item
CAS Number:361543-99-9
:
Formula:C₉H₁₃BO₃
MDL Number:MFCD03452755
:180.01 g/mol
Melting Pt:175°C
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
Specification Test Results
Related Information
Frequently Bought Together