



(+)-ɑ-Methoxy-ɑ-(trifluoromethyl)phenylacetyl chloride ∼18% ∼1.0 mol/L in dichloromethane for determination of the optical purity of alcohols and amines
Catalog # TCIAM2214-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Optical purity (GC):min. 99.0 ee%
- Concentration (Argentmetric titration):18.0 to 20.0 w/w%
Specification Test Results
About this item
:
:252.62 g/mol
Merck Index:14,06280
Storage Temperature:Refrigerator
UN:3265
ADR:8,III
:GHS05
:GHS07
:GHS08
:GHS09
CAS Number:20445-33-4
Formula:C₁₀H₈ClF₃O₂
MDL Number:MFCD00067105