Order Entry
Italy
ContactUsLinkComponent
1-Methyl-1H-indazole-3-carboxylic acid ≥98.0% (by GC, titration analysis)
Irritant
1-Methyl-1H-indazole-3-carboxylic acid ≥98.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
Irritant
1-Methyl-1H-indazole-3-carboxylic acid ≥98.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Neutralization titration):
    min. 98.0 %

Specification Test Results

Frequently Bought Together