
1-Methyl-1-propylpyrrolidinium bis(trifluoromethanesulphonyl)imide ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:223437-05-6
:
Formula:C₁₀H₁₈F₆N₂O₄S₂
MDL Number:MFCD08458917
:408.37 g/mol
Melting Pt:12°C
:GHS07
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (Ion exchange titration):min. 98.0 %
Specification Test Results
Frequently Bought Together