Order Entry
Italy
ContactUsLinkComponent
Methyl-2,5-bis(trifluoromethyl)-3,6-dioxaundecafluorononanoate (mixture of isomers) ≥95.0% (by GC)
Methyl-2,5-bis(trifluoromethyl)-3,6-dioxaundecafluorononanoate (mixture of isomers) ≥95.0% (by GC)
Catalog # TCIAM2030-5G
Supplier:  TCI
CAS Number:  
Methyl-2,5-bis(trifluoromethyl)-3,6-dioxaundecafluorononanoate (mixture of isomers) ≥95.0% (by GC)
Catalog # TCIAM2030-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 95.0 % (total of isomer)

Specification Test Results

About this item

 :
Boiling Pt:155°C
 :510.10 g/mol
CAS Number:26131-32-8
Formula:C₁₀H₃F₁₇O₄
MDL Number:MFCD00054723