
1,1-Dimethylbiguanide hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1115-70-4
:
Formula:C₄H₁₂ClN₅
MDL Number:MFCD00012582
:165.625 g/mol
Merck Index:14,05938
EINECS:214-230-6
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:219.0 to 223.0 deg-C
Specification Test Results
Frequently Bought Together