Order Entry
Italy
ContactUsLinkComponent
2-Methoxy-5-(trifluoromethoxy)benzaldehyde ≥98.0% (by GC)
Irritant
2-Methoxy-5-(trifluoromethoxy)benzaldehyde ≥98.0% (by GC)
Catalog # TCIAM1945-5G
Supplier:  TCI
CAS Number:  
Irritant
2-Methoxy-5-(trifluoromethoxy)benzaldehyde ≥98.0% (by GC)
Catalog # TCIAM1945-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Light orange to Yellow to Green clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :220.15 g/mol
 :GHS07
CAS Number:145742-65-0
Formula:C₉H₇F₃O₃
MDL Number:MFCD04115958