4-(Methylsulphonyl)phenylacetic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:90536-66-6
:
Formula:C₉H₁₀O₄S
MDL Number:MFCD00216495
:214.24 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 98.0 %
- Melting point:136.0 to 140.0 deg-C
Specification Test Results
Frequently Bought Together