
2,2'-Methylenebis(6-cyclohexyl-p-cresol) ≥97.0% (by HPLC)
TCIAM1896-25G
: TCI
:
- Pk:25 g
- Seq:3308
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 97.0 area%
- Melting point:132.0 to 136.0 deg-C
:
:392.58 g/mol
:223-773-8
:GHS07
:4066-02-8
:C₂₇H₃₆O₂