
4-Methoxy-2-(trimethylsilyl)phenyl trifluoromethanesulphonate ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:556812-41-0
:
Formula:C₁₁H₁₅F₃O₄SSi
:328.38 g/mol
UN:3265
ADR:8,II
:GHS05
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
Frequently Bought Together