Order Entry
Italy
ContactUsLinkComponent
3-Methoxy-2-(trimethylsilyl)phenyl trifluoromethanesulphonate ≥95.0% (by GC)
Corrosive
3-Methoxy-2-(trimethylsilyl)phenyl trifluoromethanesulphonate ≥95.0% (by GC)
Supplier:  TCI
Certificates
Corrosive
3-Methoxy-2-(trimethylsilyl)phenyl trifluoromethanesulphonate ≥95.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Light orange to Yellow clear liquid
  • Purity (GC):
    min. 95.0 %

Specification Test Results

Frequently Bought Together