
2-Methyl-6-(trimethylsilyl)phenyl trifluoromethanesulphonate ≥96.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:556812-44-3
:
Formula:C₁₁H₁₅F₃O₃SSi
ADR:8,II
:312.38 g/mol
UN:3265
:GHS05
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 96.0 %
Specification Test Results
Frequently Bought Together