Order Entry
Italy
ContactUsLinkComponent
4-Methyl-2-(trimethylsilyl)phenyl trifluoromethanesulphonate ≥98.0% (by GC)
Corrosive
4-Methyl-2-(trimethylsilyl)phenyl trifluoromethanesulphonate ≥98.0% (by GC)
Supplier:  TCI
Certificates
Corrosive
4-Methyl-2-(trimethylsilyl)phenyl trifluoromethanesulphonate ≥98.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

Frequently Bought Together