
Morpholinomethyl polystyrene resin 2.9 - 3.5 mmol/g, cross-linked with 1% DVB (50-100 mesh)
Catalog # TCIAM1857-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Loading:2.9 to 3.5 mmol N/g
Specification Test Results
About this item
:
:GHS07
MDL Number:MFCD00147659