
2-(4-Methyl-5-thiazolyl)ethyl acetate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:656-53-1
:
Formula:C₈H₁₁NO₂S
MDL Number:MFCD00005338
:185.24 g/mol
Boiling Pt:118°C
EINECS:211-515-7
:GHS07
Specification Test Results
- Appearance:White to Yellow to Green clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together